
1,4-Bis(acryloyl)piperazine

Synonym(s):
1,4-Di(acryloyl)piperazine, N,N′-Bis(acryloyl)piperazine, BAP, PDA, Piperazine diacrylamide
Empirical Formula (Hill Notation):
C10H14N2O2
CAS Number:
Molecular Weight:
194.23
Products
Product Category
Brand
Formula Weight
Melting Point (°C)
Physical Form
Product Line
Purity
Quality Segment
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing