Skip to Content
Merck
All Photos(1)

Key Documents

77696

Supelco

L-Kynurenine

analytical standard

Synonym(s):

β-Anthraniloyl-L-alanine, L-2-Amino-4-(2-aminophenyl)-4-oxobutanoic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H12N2O3
CAS Number:
Molecular Weight:
208.21
MDL number:
UNSPSC Code:
41116107

grade

analytical standard

Quality Level

shelf life

limited shelf life, expiry date on the label

storage temp.

2-8°C

SMILES string

N[C@@H](CC(=O)c1ccccc1N)C(O)=O

InChI

1S/C10H12N2O3/c11-7-4-2-1-3-6(7)9(13)5-8(12)10(14)15/h1-4,8H,5,11-12H2,(H,14,15)/t8-/m0/s1

InChI key

YGPSJZOEDVAXAB-QMMMGPOBSA-N

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

Key intermediate in the breakdown pathway of tryptophan.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service