Skip to Content
Merck
All Photos(1)

Key Documents

SML0574

Sigma-Aldrich

YM-298198 hydrochloride

≥98% (HPLC)

Synonym(s):

6-Amino-N-cyclohexyl-N,3-dimethyl-thiazolo[3,2-a]benzimidazole-2-carboxamide hydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C18H22N4OS · HCl
CAS Number:
Molecular Weight:
378.92
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

powder

storage condition

desiccated

color

white to beige

solubility

H2O: 5 mg/mL, clear

storage temp.

2-8°C

SMILES string

Cl.CN(C1CCCCC1)C(=O)c2sc3nc4ccc(N)cc4n3c2C

InChI

1S/C18H22N4OS.ClH/c1-11-16(17(23)21(2)13-6-4-3-5-7-13)24-18-20-14-9-8-12(19)10-15(14)22(11)18;/h8-10,13H,3-7,19H2,1-2H3;1H

InChI key

WYTJVUVCSUWZTH-UHFFFAOYSA-N

Biochem/physiol Actions

YM-298198 is an orally active, noncompetitive mGlu1 receptor antagonist. The compound inhibits glutamate-induced inositol phosphate production in NIH3T3 cells (IC50 = 16 nM). YM-298198 displays analgesic and antinociceptive properties in vivo.

Features and Benefits

This compound is featured on the Glutamate Receptors (G Protein Family) page of the Handbook of Receptor Classification and Signal Transduction. To browse other handbook pages, click here.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Haein Weon et al.
Life sciences, 188, 110-117 (2017-09-09)
Both N-type and P/Q-type voltage-gated Ca (S)-3,5-Dihydroxyphenylglycine (DHPG; 10μM for 5min), the group I mGluR agonist, was used to induce LTP of excitatory postsynaptic currents that were evoked in the Vo neurons by stimulating the trigeminal track. Weak blockade of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service