Key Documents
Z257575
Sigma-Aldrich
PROHEAT® heat gun
PH-2100, AC/DC input 220 V AC, 3 - 6 A, Schuko plug
About This Item
UNSPSC Code:
41102404
NACRES:
NB.76
material
plastic
feature
CE compliant
parameter
221-449 °C
453 L/min flow rate
output
750-1300 W
General description
Compact, lightweight heat guns with dual temperature settings of 430°F and 840°F (221°C and 449°C). Excellent source of flameless heat with air flow at 15 cfm. The PROHEAT heat guns meet most general laboratory applications.
Legal Information
PROHEAT is a registered trademark of Master Appliance Corp.