Skip to Content
Merck
All Photos(1)

Key Documents

OCNQAK490

Sigma-Aldrich

SMCC Conjugation Kit for Amine Cd-based Core/Shell Quantum Dots

fluorescence λem 490 nm, for 4 conjugation

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
43211915
NACRES:
NA.23

fluorescence

λem 490 nm

Looking for similar products? Visit Product Comparison Guide

General description

Components:
  • QSA
  • Sulfo-SMCC
  • Traut′s Reagent
  • Coupling Buffer
  • Storage Buffer
  • Quenching Buffer
  • Tubes 4 x 1.7 mL

Legal Information

Product of Ocean Nanotech, LLC.

Pictograms

Skull and crossbonesEnvironment

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Aquatic Chronic 2

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service