450170
Barium hydroxide hydrate
99.995% trace metals basis
Synonym(s):
Barium dihydroxide hydrate
About This Item
Recommended Products
Assay
99.995% trace metals basis
form
crystalline
composition
Degree of hydration, ~8
impurities
≤2.0% BaCO3 by HCl
≤55.0 ppm Trace Metal Analysis
mp
78 °C (lit.)
density
2.18 g/mL at 25 °C (lit.)
SMILES string
O.O[Ba]O
InChI
1S/Ba.3H2O/h;3*1H2/q+2;;;/p-2
InChI key
GKQTUHKAQKWLIN-UHFFFAOYSA-L
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Dam. 1 - Skin Corr. 1B
Storage Class Code
8A - Combustible corrosive hazardous materials
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service