Skip to Content
Merck
All Photos(1)

Key Documents

293849

Sigma-Aldrich

Phenylacetic acid-2-13C

99 atom % 13C

Synonym(s):

Phenylacetic acid-α-13C

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H513CH2CO2H
CAS Number:
Molecular Weight:
137.14
MDL number:
UNSPSC Code:
12352202
PubChem Substance ID:

isotopic purity

99 atom % 13C

Quality Level

form

solid

bp

265 °C (lit.)

mp

77-79 °C (lit.)

mass shift

M+1

SMILES string

OC(=O)[13CH2]c1ccccc1

InChI

1S/C8H8O2/c9-8(10)6-7-4-2-1-3-5-7/h1-5H,6H2,(H,9,10)/i6+1

InChI key

WLJVXDMOQOGPHL-PTQBSOBMSA-N

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

269.6 °F

Flash Point(C)

132.00 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

William M Moe et al.
Environmental science & technology, 52(16), 9311-9319 (2018-07-26)
In situ bioremediation practices that include subsurface addition of fermentable electron donors to stimulate reductive dechlorination by anaerobic bacteria have become widely employed to combat chlorinated solvent contamination in groundwater. At a contaminated site located near Baton Rouge, Louisiana (USA)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service