TLC Analysis of Nucleosides on HPTLC Silica gel 60 RP-18 WF254s
Materials
standard
Product No.
Description
Pricing
CONDITIONS
plate
HPTLC pre-coated plate RP-18 WF 254s with concentrating zone (1.13124.0001)
eluent
[A] water; [B] ethanol; (95:5, A:B)
migration distance
7 cm
chamber
normal chamber without chamber saturation
sample
5 μL (0.005%)
detector
in situ evaluation with TLC/HPTLC scanner (Camag) UV 265 nm