Merck
Shell Thickness Engineering Significantly Boosts the Photocatalytic H
ACS applied materials & interfaces (2017-09-28)
Ping Wang, Minmin Wang, Jie Zhang, Chuanping Li, Xiaolong Xu, Yongdong Jin
ABSTRACT
Colloidal semiconductor quantum dots (QDs) have recently emerged as a good candidate for photocatalytic hydrogen (H
Sigma-Aldrich
Sulfur, purum p.a., ≥99% (T)