Merck
  • An efficient enzyme-powered micromotor device fabricated by cyclic alternate hybridization assembly for DNA detection.
An efficient enzyme-powered micromotor device fabricated by cyclic alternate hybridization assembly for DNA detection.
Nanoscale (2017-06-24)
Shizhe Fu, Xueqing Zhang, Yuzhe Xie, Jie Wu, Huangxian Ju
ABSTRACT
An efficient enzyme-powered micromotor device was fabricated by assembling multiple layers of catalase on the inner surface of a poly(3,4-ethylenedioxythiophene and sodium 4-styrenesulfonate)/Au microtube (PEDOT-PSS/Au). The catalase assembly was achieved by programmed DNA hybridization, which was performed by immobilizing a designed sandwich DNA structure as the sensing unit on the PEDOT-PSS/Au, and then alternately hybridizing with two assisting DNA to bind the enzyme for efficient motor motion. The micromotor device showed unique features of good reproducibility, stability and motion performance. Under optimal conditions, it showed a speed of 420 μm s
Sigma-Aldrich
Sodium 4-vinylbenzenesulfonate, technical, ≥90% (T)
Sigma-Aldrich
3,4-Ethylenedioxythiophene, 97%