Merck
  • HMGB1 induces human non-small cell lung cancer cell motility by activating integrin αvβ3/FAK through TLR4/NF-κB signaling pathway.
HMGB1 induces human non-small cell lung cancer cell motility by activating integrin αvβ3/FAK through TLR4/NF-κB signaling pathway.
Biochemical and biophysical research communications (2016-10-23)
Jianhua Zhu, Jing Luo, Yongchao Li, Min Jia, Yueqin Wang, Yan Huang, Shuhong Ke
ABSTRACT
High mobility group box 1 (HMGB1) is a ubiquitous nuclear protein with multi-functions and plays an important role in tumorigenesis and metastasis in various human cancers. In the present study, we found that HMGB1 induced migration of in human non-small cell lung cancer (NSCLC) cells by up-regulating integrin αvβ3 expression. Further investigation evidenced that HMGB1 activated Toll-like receptor 4 (TLR4) and NF-κB, which was responsible for αvβ3 up-regulation. Furthermore, HMGB1-induced integrin αvβ3 expression led to focal adhesion kinase (FAK) phosphorylation and increased paxillin and talin mRNA expression. Knockdown HMGB1 inhibited xenograft tumor metastasis in athymic mice. Taken together, our findings suggested that HMGB1 enhances tumor cell migration ability by activating αvβ3/FAK through TLR4/NF-κB signaling, leading to metastasis of NSCLC.
Sigma-Aldrich
EVP4593, ≥98% (HPLC)