Merck
  • B-Myb switches from Cyclin/Cdk-dependent to Jnk- and p38 kinase-dependent phosphorylation and associates with SC35 bodies after UV stress.
B-Myb switches from Cyclin/Cdk-dependent to Jnk- and p38 kinase-dependent phosphorylation and associates with SC35 bodies after UV stress.
Cell death & disease (2013-03-02)
E Werwein, M Dzuganova, C Usadel, K-H Klempnauer
ABSTRACT
B-Myb is a highly conserved member of the Myb transcription factor family that has essential roles in cell-cycle progression. Recent work has suggested that B-Myb is also involved in the cellular DNA-damage response. Here, we have investigated the fate of B-Myb in UV-irradiated cells. UV stress leads to the appearance of phosphorylated B-Myb in nuclear SC35 speckles during transcriptional shutdown. Furthermore, we show that UV irradiation leads to a change of the phosphorylation pattern of B-Myb, which is caused by a switch from Cyclin/Cdk-dependent to Jnk and p38 kinase-dependent phosphorylation. Taken together, we have identified Jnk and p38 kinase as novel regulators of B-Myb and established the localization of phosphorylated B-Myb in SC35 speckles as a potential novel regulatory mechanism for B-Myb in UV irradiated cells.
Sigma-Aldrich
Anti-SC-35 Antibody, clone 1SC-4F11, ascites fluid, clone 1SC-4F11, from mouse
Sign Into View Organizational & Contract Pricing
SKUVendor SKUPack SizeAvailabilityPriceQuantity
EN300-179218-100MG100 mg
Ships in 14 days - from Aldrich Partner.
€380.40
EN300-179218-250MG250 mg
Ships in 14 days - from Aldrich Partner.
€527.30
EN300-179218-500MG500 mg
Ships in 14 days - from Aldrich Partner.
€802.90
EN300-179218-1G1 g
Ships in 14 days - from Aldrich Partner.
€1,017.40
Sigma-Aldrich
Anti-HET/SAF-B Antibody, clone 6F7, clone 6F7, Upstate®, from mouse
Sign Into View Organizational & Contract Pricing
SKUVendor SKUPack SizeAvailabilityPriceQuantity
EN300-179218-100MG100 mg
Ships in 14 days - from Aldrich Partner.
€380.40
EN300-179218-250MG250 mg
Ships in 14 days - from Aldrich Partner.
€527.30
EN300-179218-500MG500 mg
Ships in 14 days - from Aldrich Partner.
€802.90
EN300-179218-1G1 g
Ships in 14 days - from Aldrich Partner.
€1,017.40