Merck
  • Systematic review: effective management strategies for lactose intolerance.
Systematic review: effective management strategies for lactose intolerance.
Annals of internal medicine (2010-04-21)
Aasma Shaukat, Michael D Levitt, Brent C Taylor, Roderick MacDonald, Tatyana A Shamliyan, Robert L Kane, Timothy J Wilt
ABSTRACT
Lactose intolerance resulting in gastrointestinal symptoms is a common health concern. Diagnosis and management of this condition remain unclear. To assess the maximum tolerable dose of lactose and interventions for reducing symptoms of lactose intolerance among persons with lactose intolerance and malabsorption. Multiple electronic databases, including MEDLINE and the Cochrane Library, for trials published in English from 1967 through November 2009. Randomized, controlled trials of individuals with lactose intolerance or malabsorption. Three investigators independently reviewed articles, extracted data, and assessed study quality. 36 unique randomized studies (26 on lactase- or lactose-hydrolyzed milk supplements, lactose-reduced milk, or tolerable doses of lactose; 7 on probiotics; 2 on incremental lactose administration for colonic adaptation; and 1 on another agent) met inclusion criteria. Moderate-quality evidence indicated that 12 to 15 g of lactose (approximately 1 cup of milk) is well tolerated by most adults. Evidence was insufficient that lactose-reduced solution or milk with a lactose content of 0 to 2 g, compared with greater than 12 g, is effective in reducing symptoms of lactose intolerance. Evidence for probiotics, colonic adaptation, and other agents was also insufficient. Most studies evaluated persons with lactose malabsorption rather than lactose intolerance. Variation in enrollment criteria, outcome reporting, and the composition and dosing of studied agents precluded pooling of results and limited interpretation. Most individuals with presumed lactose intolerance or malabsorption can tolerate 12 to 15 g of lactose. Additional studies are needed to determine the effectiveness of lactose intolerance treatment.
Product Description
Supelco
Lactose, Anhydrous, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Lactose, tested according to Ph. Eur.
Lactose (anhydrous), European Pharmacopoeia (EP) Reference Standard
USP
Anhydrous lactose, United States Pharmacopeia (USP) Reference Standard