Merck
  • Mechanism of volatile compound production during storage of sunflower Oil.
Mechanism of volatile compound production during storage of sunflower Oil.
Journal of agricultural and food chemistry (2001-01-16)
A Keszler, T Kriska, A Németh
ABSTRACT
Volatile compounds formed in the course of the thermal decomposition of hydroperoxides during storage of sunflower oil were analyzed by headspace solid-phase microextraction sampling followed by gas chromatographic separation and mass spectral detection. The determining role of alkoxyl radicals in the process has been proven by electron spin resonance spectroscopic measurements. On the basis of analytical results, the reaction networks and mechanisms were constructed by computer modeling to describe the formation of volatile products of radical decomposition of hydroperoxides. We established that off-flavor aliphatic aldehydes are originated from only the alkoxyl radicals derived from trigliceride of linoleic acid. To find a specific additive, which redirects the formation of these radicals toward production of more stable species, is suggested.
Product Description
Supelco
SPME fiber assembly Polydimethylsiloxane (PDMS), metal alloy fiber, df 100 μm(PDMS, needle size 23 ga, for use with autosampler
Supelco
SPME fiber assembly Polydimethylsiloxane (PDMS), df 100 μm(PDMS, for use with autosampler, needle size 24 ga
Supelco
SPME Portable Field Sampler, coating PDMS
Supelco
SPME fiber assembly Polydimethylsiloxane (PDMS), df 100 μm(PDMS, for use with manual holder, needle size 23 ga
Supelco
SPME fiber assembly Polydimethylsiloxane (PDMS), df 100 μm(PDMS, for use with autosampler, needle size 23 ga
Supelco
SPME fiber assembly Polydimethylsiloxane (PDMS), df 100 μm(PDMS, needle size 24 ga, for use with manual holder