Merck
  • Chemical composition and biological activity of the essential oil from leaves of Moringa oleifera Lam. cultivated in Mozambique.
Chemical composition and biological activity of the essential oil from leaves of Moringa oleifera Lam. cultivated in Mozambique.
Molecules (Basel, Switzerland) (2013-09-12)
Tatiana Marrufo, Filomena Nazzaro, Emilia Mancini, Florinda Fratianni, Raffaele Coppola, Laura De Martino, Adelaide Bela Agostinho, Vincenzo De Feo
ABSTRACT
The antioxidant capacity and antimicrobial activity of the essential oil of Moringa oleifera (Moringaceae) grown in Mozambique was investigated. The chemical composition was studied by means of GC and GC-MS analysis. Hexacosane (13.9%), pentacosane (13.3%) and heptacosane (11.4%) were the main components. Ultra High Performance Chromatography-DAD analysis detected the flavonoids quercetin (126 μg/g) and luteolin (6.2 μg/g). The essential oil exhibited a relatively low free radical scavenging capacity. The antimicrobial activity of the essential oil was assayed against two Gram-positive strains (Bacillus cereus, Staphylococcus aureus), two Gram-negative strains (Escherichia coli, Pseudomonas aeruginosa), and five fungal strains of agro-food interest (Penicillium aurantiogriseum, Penicillium expansum, Penicillium citrinum, Penicillium digitatum, and Aspergillus niger spp.). B. cereus and P. aeruginosa, as well as the fungal strains were sensitive to the essential oil.
Sigma-Aldrich
Hexacosane, 99%