Merck
  • Gelatin as a delivery vehicle for the controlled release of bioactive molecules.
Gelatin as a delivery vehicle for the controlled release of bioactive molecules.
Journal of controlled release : official journal of the Controlled Release Society (2005-11-04)
Simon Young, Mark Wong, Yasuhiko Tabata, Antonios G Mikos
ABSTRACT
Gelatin is a commonly used natural polymer which is derived from collagen. The isoelectric point of gelatin can be modified during the fabrication process to yield either a negatively charged acidic gelatin, or a positively charged basic gelatin at physiological pH. This theoretically allows electrostatic interactions to take place between a charged biomolecule and gelatin of the opposite charge, forming polyion complexes. Various forms of gelatin carrier matrices can be fabricated for controlled-release studies, and characterization studies have been performed which show that gelatin carriers are able to sorb charged biomolecules such as proteins and plasmid DNA through polyion complexation. The crosslinking density of gelatin hydrogels has been shown to affect their degradation rate in vivo, and the rate of biomolecule release from gelatin carriers has been shown to have a similar profile, suggesting that complexed gelatin/biomolecule fragments are released by enzymatic degradation of the carrier in vivo. This review will emphasize how biomolecules released from gelatin controlled-release systems are able to retain their biological activity, allowing for their use in tissue engineering, therapeutic angiogenesis, gene therapy, and drug delivery applications.
Sigma-Aldrich
Gelatin from porcine skin, Type A, lyophilized powder, γ-irradiated, BioXtra, suitable for cell culture
Sigma-Aldrich
Gelatin from porcine skin, powder, gel strength ~300 g Bloom, Type A, BioReagent, suitable for electrophoresis, suitable for cell culture
Sigma-Aldrich
Gelatin from bovine skin, Type B
Sigma-Aldrich
Gelatin from cold water fish skin, 40-50% in H2O
Sigma-Aldrich
Gelatin, tested according to Ph. Eur.
Sigma-Aldrich
Gelatin from bovine skin, gel strength ~225 g Bloom, Type B
Sigma-Aldrich
Prionex® Highly purified Type A, aqueous solution
Sigma-Aldrich
Gelatin from cold water fish skin, solid
Sigma-Aldrich
Gelatin from porcine skin, gel strength 300, Type A
Sigma-Aldrich
Gelatin solution, Type B, 2% in H2O, tissue culture grade, BioReagent, suitable for cell culture
Millipore
Gelatin from porcine skin, suitable for microbiology, low gel strength
Millipore
Gelatin from porcine skin, suitable for microbiology, ultrahigh gel strength
Millipore
Gelatin from porcine skin, suitable for microbiology, high gel strength
Sigma-Aldrich
Gelatin from porcine skin, gel strength ~175 g Bloom, Type A
Millipore
Gelatin from porcine skin, medium gel strength, suitable for microbiology
Sigma-Aldrich
Gelatin from porcine skin, gel strength 80-120 g Bloom, Type A
Sigma-Aldrich
Gelatin from bovine skin, Type B, powder, BioReagent, suitable for cell culture