Merck
  • Measuring environmental phenols and chlorinated organic chemicals in breast milk using automated on-line column-switching-high performance liquid chromatography-isotope dilution tandem mass spectrometry.
Measuring environmental phenols and chlorinated organic chemicals in breast milk using automated on-line column-switching-high performance liquid chromatography-isotope dilution tandem mass spectrometry.
Journal of chromatography. B, Analytical technologies in the biomedical and life sciences (2005-12-27)
Xiaoyun Ye, Zsuzsanna Kuklenyik, Larry L Needham, Antonia M Calafat
ABSTRACT
Breast milk is one possible route of exposure to environmental chemicals, including phenols and chlorinated organic chemicals for breast-fed infants. We developed a highly sensitive method of analyzing breast milk for triclocarban (3,4,4'-trichlorocarbanilide) and eight phenolic compounds: bisphenol A (BPA), 4-tert-octylphenol (4-tOP), ortho-phenylphenol (OPP), 2,4-dichlorophenol, 2,5-dichlorophenol, 2,4,5-trichlorophenol, 2,4,6-trichlorophenol, and 2-hydroxy-4-metoxybenzophenone (BP-3). The method includes adding a solution containing a stable isotope of each chemical, enzymatic hydrolysis of the conjugated chemicals in the milk, and on-line solid-phase extraction coupled with high performance liquid chromatography-tandem mass spectrometry. It can also be used to measure the free (unconjugated) species by omitting the enzymatic deconjugation step. The method, validated using pooled breast milk samples, has inter-day coefficient of variations ranging from 4.8 to 18.9% for most analytes, and spiked recoveries generally about 100%. Detection limits for most analytes are below 1 ng/mL in 100 microL of breast milk. We tested the usefulness of the method by measuring concentrations of these nine compounds in 20 breast milk samples. BPA, OPP, and BP-3 were detected in more than 60% of the samples tested. The free species of these compounds appear to be most prevalent in milk.
Sigma-Aldrich
3,4,4′-Trichlorocarbanilide, 99%
Sigma-Aldrich
2,4-Dichlorophenol, 99%