Merck
  • Feasibility of capillary liquid chromatography/microchip atmospheric pressure photoionization mass spectrometry in analyzing anabolic steroids in urine samples.
Feasibility of capillary liquid chromatography/microchip atmospheric pressure photoionization mass spectrometry in analyzing anabolic steroids in urine samples.
Rapid communications in mass spectrometry : RCM (2010-03-09)
Linda L Ahonen, Markus Haapala, Ville Saarela, Sami Franssila, Tapio Kotiaho, Risto Kostiainen
ABSTRACT
We examined the feasibility of capillary liquid chromatography/microchip atmospheric pressure photoionization tandem mass spectrometry (capLC/microAPPI-MS/MS) for the analysis of anabolic steroids in human urine. The urine samples were pretreated by enzymatic hydrolysis (with beta-glucuronidase from Helix pomatia), and the compounds were liquid-liquid extracted with diethyl ether. After separation the compounds were vaporized by microchip APPI, photoionized by a 10 eV krypton discharge lamp, and detected by selected reaction monitoring. The capLC/microAPPI-MS/MS method showed good sensitivity with detection limits at the level of 1.0 ng mL(-1), good linearity with correlation coefficients between 0.9954 and 0.9990, and good repeatability with relative standard deviations below 10%. These results demonstrate that microchip APPI combined with capLC/MS/MS provides a new potential method for analyzing non-polar and neutral compounds in biological samples.
Product Description
Supelco
17β-Hydroxy-17-methylandrosta-1,4-dien-3-one, analytical standard
Supelco
Methandienone solution, 1.0 mg/mL in 1,2-dimethoxyethane, ampule of 1 mL, certified reference material, Cerilliant®