Merck
  • Biogeochemical reductive release of soil embedded arsenate around a crater area (Guandu) in northern Taiwan using X-ray absorption near-edge spectroscopy.
Biogeochemical reductive release of soil embedded arsenate around a crater area (Guandu) in northern Taiwan using X-ray absorption near-edge spectroscopy.
Journal of environmental sciences (China) (2013-08-08)
Kai-Ying Chiang, Tsan-Yao Chen, Chih-Hao Lee, Tsang-Lang Lin, Ming-Kuang Wang, Ling-Yun Jang, Jyh-Fu Lee
ABSTRACT
This study investigates biogeochemical reductive release of arsenate from beudantite into solution in a crater area in northern Taiwan, using a combination of X-ray absorption near-edge structure (XANES) and atomic absorption spectrometry. Total arsenic (As) concentrations in the soil were more than 200 mg/kg. Over four months of laboratory experiments, less than 0.8% As was released into solution after reduction experiments. The 71% to 83% As was chemically reduced into arsenite (As(III)) and partially weathering into the soluble phase. The kinetic dissolution and re-precipitation of As, Fe, Pb and sulfate in this area of paddy soils merits further study.
Sigma-Aldrich
Lead, powder, −325 mesh, ≥99% trace metals basis
Sigma-Aldrich
Lead, shot, 1-3 mm, 99.995% trace metals basis
Sigma-Aldrich
Lead, shot, <2 mm, 99.9% trace metals basis
Sigma-Aldrich
Lead, powder, −100 mesh, 99.95% trace metals basis