Merck
  • Facile one-pot synthesis of novel dispirooxindole-pyrrolidine derivatives and their antimicrobial and anticancer activity against A549 human lung adenocarcinoma cancer cell line.
Facile one-pot synthesis of novel dispirooxindole-pyrrolidine derivatives and their antimicrobial and anticancer activity against A549 human lung adenocarcinoma cancer cell line.
Bioorganic & medicinal chemistry letters (2013-02-12)
Y Arun, G Bhaskar, C Balachandran, S Ignacimuthu, P T Perumal
ABSTRACT
Novel dispirooxindole-pyrrolidine derivatives have been synthesized through 1,3-dipolar cycloaddition of an azomethine ylide generated from isatin and sarcosine with the dipolarophile 3-(1H-indol-3-yl)-3-oxo-2-(2-oxoindolin-3-ylidene)propanenitrile, and also spiro compound of acenaphthenequinone obtained by the same optimized reaction condition. Synthesized compounds were evaluated for their antimicrobial activity and all the compounds shown significant activity. Anticancer activity was evaluated against A549 human lung adenocarcinoma cancer cell lines. Compounds 7b, 7g, 7i and 7r exhibit very good anticancer activity 62.96%, 62.03%, 67.67% and 60.22%, respectively, at the dose of 200μg/mL and compound 7i shows IC50 value in 50μg/mL.
Sigma-Aldrich
Pyrrolidine, FG
Sigma-Aldrich
Pyrrolidine, ≥99.0%
Sigma-Aldrich
Pyrrolidine, 99%
Sigma-Aldrich
Pyrrolidine, ≥99.5%, purified by redistillation