Merck
  • Probing the orientational distribution of dyes in membranes through multiphoton microscopy.
Probing the orientational distribution of dyes in membranes through multiphoton microscopy.
Biophysical journal (2012-09-27)
James E Reeve, Alex D Corbett, Igor Boczarow, Tony Wilson, Hagan Bayley, Harry L Anderson
ABSTRACT
Numerous dyes are available or under development for probing the structural and functional properties of biological membranes. Exogenous chromophores adopt a range of orientations when bound to membranes, which have a drastic effect on their biophysical behavior. Here, we present a method that employs optical anisotropy data from three polarization-imaging techniques to establish the distribution of orientations adopted by molecules in monolayers and bilayers. The resulting probability density functions, which contain the preferred molecular tilt Ī¼ and distribution breadth Ī³, are more informative than an average tilt angle [Ļ†]. We describe a methodology for the extraction of anisotropy data through an image-processing technology that decreases the error in polarization measurements by about a factor of four. We use this technique to compare di-4-ANEPPS and di-8-ANEPPS, both dipolar dyes, using data from polarized 1-photon, 2-photon fluorescence and second-harmonic generation imaging. We find that di-8-ANEPPS has a lower tilt but the same distributional width. We find the distribution of tilts taken by di-4-ANEPPS in two phospholipid membrane models: giant unilamellar vesicles and water-in-oil droplet monolayers. Both models result in similar distribution functions with average tilts of 52Ā° and 47Ā°, respectively.
Sigma-Aldrich
4-(2-(6-(Dibutylamino)-2-naphthalenyl)ethenyl)-1-(3-sulfopropyl)pyridinium hydroxide inner salt, ≥95% (HPLC), solid