Merck
  • Caged fluorescent haptens reveal the generation of cryptic epitopes in allergic contact dermatitis.
Caged fluorescent haptens reveal the generation of cryptic epitopes in allergic contact dermatitis.
The Journal of investigative dermatology (2011-01-14)
Carl Simonsson, Sofia I Andersson, Anna-Lena Stenfeldt, Jörgen Bergström, Brigitte Bauer, Charlotte A Jonsson, Marica B Ericson, Kerstin S Broo
ABSTRACT
Allergic contact dermatitis (ACD) is the most prevalent form of human immunotoxicity. It is caused by skin exposure to haptens, i.e., protein-reactive, low-molecular-weight chemical compounds, which form hapten-protein complexes (HPCs) in the skin, triggering the immune system. These immunogenic HPCs are elusive. In this study a series of thiol-reactive caged fluorescent haptens, i.e., bromobimanes, were deployed in combination with two-photon fluorescence microscopy, immunohistochemistry, and proteomics to identify possible hapten targets in proteins in human skin. Key targets found were the basal keratinocytes and the keratins K5 and K14. Particularly, cysteine 54 of K5 was found to be haptenated by the bromobimanes. In addition, elevated levels of anti-keratin antibodies were found in the sera of mice exposed to bromobimanes in vivo. The results indicate a general mechanism in which thiol-reactive haptens generate cryptic epitopes normally concealed from the immune system. In addition, keratinocytes and keratin seem to have an important role in the mechanism behind ACD, which is a subject for further investigations.
Sigma-Aldrich
Bromobimane, suitable for fluorescence, BioReagent, ≥95% (HPCE)
Sigma-Aldrich
Bromobimane, ≥97% (HPLC)