Merck
  • Synthesis of substituted naphthalenes via a catalytic ring-expansion rearrangement.
Synthesis of substituted naphthalenes via a catalytic ring-expansion rearrangement.
Organic letters (2008-10-07)
Adam C Glass, Benjamin B Morris, Lev N Zakharov, Shih-Yuan Liu
ABSTRACT
A new methodology for the preparation of substituted naphthalenes starting from readily available indenones, organometal reagents, and trimethylsilyldiazomethane via a catalytic rearrangement process is described. Hindered biaryl naphthalenes, including triortho-substituted biaryls, can be accessed through our method. Our results are consistent with a mechanism involving a benzobenzvalene intermediate.
Sigma-Aldrich
Indene, ≥99%
Sigma-Aldrich
Indene, 98%
Sigma-Aldrich
Indene, contains 50-100 ppm tert-butylcatechol as stabilizer, technical grade, ≥90%