Merck
  • Formation of gamma-lactones through CAN-mediated oxidative cleavage of hemiketals.
Formation of gamma-lactones through CAN-mediated oxidative cleavage of hemiketals.
The Journal of organic chemistry (2008-08-13)
Alexander M Jacobine, Weimin Lin, Bethany Walls, Charles K Zercher
ABSTRACT
The generation of substituted gamma-lactones can be accomplished through application of a tandem chain extension-aldol reaction, followed by CAN-mediated oxidative cleavage of the aldol product. The oxidative cleavage requires the intermediacy of a hemiketal and the presence of an alpha-heteroatom. Formation of the gamma-lactone through the oxidative cleavage is used to assign stereochemistry of the aldol reaction and as the final step in a short synthesis of members of the phaseolinic acid family of natural products.
Sigma-Aldrich
Ammonium cerium(IV) nitrate, ≥99.99% trace metals basis
Sigma-Aldrich
Ammonium cerium(IV) nitrate, ACS reagent, ≥98.5%
Sigma-Aldrich
Ammonium cerium(IV) nitrate, puriss. p.a., ACS reagent, ≥98.5% (RT)
Sigma-Aldrich
Ammonium cerium(IV) nitrate, ≥98% (titration)