Merck
  • Evidence of heterotrimeric G-protein involvement in regulated exocytosis from permeabilized pancreatic acini.
Evidence of heterotrimeric G-protein involvement in regulated exocytosis from permeabilized pancreatic acini.
Pancreas (1995-05-01)
R C De Lisle, G W Howell
ABSTRACT
Constitutive membrane trafficking events are regulated by heterotrimeric G-proteins (G-proteins) in addition to their regulation by small GTP-binding proteins (smgs). Here, we used streptolysin O-permeabilized mouse pancreatic acini and compounds that interact with G-proteins, but not smgs, to examine whether G-proteins are also involved in regulated pancreatic exocytosis. The wasp venom mastoparan (10 microM) inhibited by 25-50% amylase release from permeabilized acini stimulated by various combinations of Ca2+, cyclic AMP (cAMP), 12-O-tetradecanoylphorbol 13-acetate, and guanosine (5'-[gamma-thio]triphosphate (GTP gamma S), while the inactive analogue Mas17 was without effect. Pretreatment of intact acini with pertussis toxin resulted in an approximately 30% reduction of amylase secretion from cells subsequently permeabilized and stimulated with calcium and GTP gamma S. Pretreatment of intact acini with cholera toxin increased stimulated amylase release by 30% from subsequently permeabilized cells, and this effect was mimicked by 8-Br-cAMP. The cAMP-dependent protein kinase inhibitor H-89 (3 microM) largely reversed the effect of cholera toxin, indicating that cholera toxin's effect is due to increased cellular cAMP levels. The inhibitory effects of mastoparan and pertussis toxin suggest that a Gi/Go-type G-protein(s) is (are) involved in the regulation of exocytosis. Since mastoparan inhibited exocytosis stimulated by all intracellular mediators tested, it indicates that the G-protein acts at a distal step in the exocytic process.
Sigma-Aldrich
Mastoparan 17, ≥96.5%, lyophilized powder
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
100 μL
1 Estimated to ship on 03 March 2025
A$1,040.00