Merck
  • The natural course of gross motor deterioration in metachromatic leukodystrophy.
The natural course of gross motor deterioration in metachromatic leukodystrophy.
Developmental medicine and child neurology (2011-06-29)
Christiane Kehrer, Gunnar Blumenstock, Volkmar Gieselmann, Ingeborg Krägeloh-Mann
ABSTRACT
Motor deterioration is a key feature in metachromatic leukodystrophy (MLD). The lack of data about its natural course impedes evaluation of therapeutic interventions. This study aimed to provide data about motor decline in MLD. Fifty-nine patients (27 males, 32 females) with MLD (21 with late-infantile MLD and 38 with juvenile MLD) were recruited within a nationwide survey (the German LEUKONET). Median (range) age at onset was 17 months (9-27) for the group with late-infantile MLD and 6 years 2 months (2y 11mo-14y) for the group with juvenile MLD. Gross motor function was assessed using the Gross Motor Function Classification for MLD. In late-infantile MLD, all patients showed loss of all gross motor function until 3 years 4 months of age. Patients with juvenile MLD showed a more variable and significantly longer motor decline (p<0.001). For a patient with the juvenile form showing first gait disturbances, the probability of remaining stable for more than 1 year was 84%, and 51% for more than 2 years. Having lost independent walking, subsequent motor decline was as steep as in the late-infantile form (median 5 mo, interquartile range 3-22). The course of motor disease was more variable in juvenile MLD with respect to onset and dynamics. However, the motor decline after the loss of independent walking was similarly steep in both forms. These data can serve as a reference for clinical studies that are topics of current research and allow definition of inclusion/exclusion criteria.
Sigma-Aldrich
Sulfatase from Aerobacter aerogenes, Type VI, buffered aqueous glycerol solution, 2-5 units/mg protein (biuret), 10-20 units/mL
Sigma-Aldrich
Sulfatase from abalone entrails, Type VIII, lyophilized powder, 20-40 units/mg solid
Sigma-Aldrich
Sulfatase from Patella vulgata (keyhole limpet), Type IV, essentially salt-free, lyophilized powder, ≥10 units/mg solid
Sigma-Aldrich
Sulfatase from Helix pomatia, Type H-1, sulfatase ≥10,000 units/g solid
Sigma-Aldrich
Sulfatase from Helix pomatia, Type H-2, aqueous solution, ≥2,000 units/mL