Merck
  • Tristetraprolin Posttranscriptionally Downregulates TRAIL Death Receptors.
Tristetraprolin Posttranscriptionally Downregulates TRAIL Death Receptors.
Cells (2020-08-14)
Won Hyeok Lee, Myung Woul Han, Song Hee Kim, Daseul Seong, Jae Hee An, Hyo Won Chang, Sang Yoon Kim, Seong Who Kim, Jong Cheol Lee
ABSTRACT
Tumor necrosis factor (TNF)-related apoptosis-inducing ligand (TRAIL) has attracted attention as a potential candidate for cancer therapy. However, many primary cancers are resistant to TRAIL, even when combined with standard chemotherapy. The mechanism of TRAIL resistance in cancer cells has not been fully elucidated. The TRAIL death receptor (DR) 3'-untranslated region (3'-UTR) is reported to contain AU-rich elements (AREs) that are important for regulating DR mRNA stability. However, the mechanisms by which DR mRNA stability is determined by its 3'-UTR are unknown. We demonstrate that tristetraprolin (TTP), an ARE-binding protein, has a critical function of regulating DR mRNA stability. DR4 mRNA contains three AREs and DR5 mRNA contains four AREs in 3'-UTR. TTP bound to all three AREs in DR4 and ARE3 in DR5 and enhanced decay of DR4/5 mRNA. TTP overexpression in colon cancer cells changed the TRAIL-sensitive cancer cells to TRAIL-resistant cells, and down-regulation of TTP increased TRAIL sensitivity via DR4/5 expression. Therefore, this study provides a molecular mechanism for enhanced levels of TRAIL DRs in cancer cells and a biological basis for posttranscriptional modification of TRAIL DRs. In addition, TTP status might be a biomarker for predicting TRAIL response when a TRAIL-based treatment is used for cancer.
Product Description
Supelco
EPA 8260A Surrogate Standards Mix, certified reference material, 2000 μg/mL each component in methanol
Sigma-Aldrich
Thymidine 5′-triphosphate sodium salt, ≥96%
Sigma-Aldrich
IgG from mouse serum, reagent grade, ≥95% (SDS-PAGE), lyophilized powder