Merck
  • A Conserved Proline Hinge Mediates Helix Dynamics and Activation of Rhodopsin.
A Conserved Proline Hinge Mediates Helix Dynamics and Activation of Rhodopsin.
Structure (London, England : 1993) (2020-05-30)
Andreyah L Pope, Omar B Sanchez-Reyes, Kieron South, Ekaterina Zaitseva, Martine Ziliox, Reiner Vogel, Philip J Reeves, Steven O Smith
ABSTRACT
Despite high-resolution crystal structures of both inactive and active G protein-coupled receptors (GPCRs), it is still not known how ligands trigger the large structural change on the intracellular side of the receptor since the conformational changes that occur within the extracellular ligand-binding region upon activation are subtle. Here, we use solid-state NMR and Fourier transform infrared spectroscopy on rhodopsin to show that Trp2656.48 within the CWxP motif on transmembrane helix H6 constrains a proline hinge in the inactive state, suggesting that activation results in unraveling of the H6 backbone within this motif, a local change in dynamics that allows helix H6 to swing outward. Notably, Tyr3017.48 within activation switch 2 appears to mimic the negative allosteric sodium ion found in other family A GPCRs, a finding that is broadly relevant to the mechanism of receptor activation.
Sigma-Aldrich
Anti-Rhodopsin Antibody, CT, last 9 amino acids, clone Rho 1D4, clone Rho 1D4, Chemicon®, from mouse
Sigma-Aldrich
Benzamidine, ≥95.0%
Sigma-Aldrich
Phenol-4-13C, 99 atom % 13C
Sigma-Aldrich
Tetracycline hydrochloride, powder