Merck
Activation of the Unfolded Protein Response by Lipid Bilayer Stress.
Molecular cell (2017-07-12)
Kristina Halbleib, Kristina Pesek, Roberto Covino, Harald F Hofbauer, Dorith Wunnicke, Inga Hänelt, Gerhard Hummer, Robert Ernst
ABSTRACT
The unfolded protein response (UPR) is a conserved homeostatic program that is activated by misfolded proteins in the lumen of the endoplasmic reticulum (ER). Recently, it became evident that aberrant lipid compositions of the ER membrane, referred to as lipid bilayer stress, are equally potent in activating the UPR. The underlying molecular mechanism, however, remained unclear. We show that the most conserved transducer of ER stress, Ire1, uses an amphipathic helix (AH) to sense membrane aberrancies and control UPR activity. In vivo and in vitro experiments, together with molecular dynamics (MD) simulations, identify the physicochemical properties of the membrane environment that control Ire1 oligomerization. This work establishes the molecular mechanism of UPR activation by lipid bilayer stress.
Product Description
Roche
Anti-HA High Affinity, from rat IgG1
Sigma-Aldrich
Monoclonal ANTI-FLAG® M2 antibody produced in mouse, clone M2, purified immunoglobulin (Purified IgG1 subclass), buffered aqueous solution (10 mM sodium phosphate, 150 mM NaCl, pH 7.4, containing 0.02% sodium azide)