Merck
  • PEGylation and surface functionalization of liposomes containing drug nanocrystals for cell-targeted delivery.
PEGylation and surface functionalization of liposomes containing drug nanocrystals for cell-targeted delivery.
Colloids and surfaces. B, Biointerfaces (2019-07-28)
Yunxin Xiao, Qingtao Liu, Andrew J Clulow, Tang Li, Madhura Manohar, Elliot P Gilbert, Liliana de Campo, Adrian Hawley, Ben J Boyd
ABSTRACT
Liposomal formulations have important therapeutic applications in anti-cancer treatments but current formulations suffer from serious side effects, high dosage requirements and prolonged treatment. In this study, PEGylated azide-functionalized liposomes containing drug nanocrystals were investigated with the aim of increasing the drug payload and achieving functionalization for targeted delivery. Liposomes were characterized using cryogenic transmission electron microscopy (cryo-TEM), dynamic light scattering (DLS), small and ultra-small angle neutron scattering (SANS/USANS) and small and wide angle X-ray scattering (SAXS/WAXS). Cryo-TEM experiments revealed the dimensions of the nanocrystal-loaded liposomes and the change of shape from spherical to elongated after the formation of nanocrystals. Results from SANS/USANS experiments confirmed the asymmetric particle shape. SAXS/WAXS experiments confirmed that the crystalline drug only occurred in freeze-thawed samples and correlated with a new unidentified polymorphic form of ciprofloxacin. Using a small molecule dye, dibenzocyclooctyne (DBCO)-cy5, specific conjugation between DBCO groups and surface azide groups on the liposomes was confirmed; this indicates the promise of this system for tumour-targeted delivery.
Sigma-Aldrich
1,2-Distearoyl-sn-glycero-3-phosphoethanolamine, ≥99%
Sigma-Aldrich
NanoFabTx - Azide Lipid Mix, for synthesis of azide-functionalized liposomes
Avanti
DSPE-PEG(2000) Azide, Avanti Research - A Croda Brand 880228C
Sigma-Aldrich
Triton X-100, laboratory grade