
Ziprasidone hydrochloride solution

Empirical Formula (Hill Notation):
C21H21ClN4OS · HCl
CAS Number:
Molecular Weight:
449.40
Products
Brand
Formula Weight
Manufacturer
Format
Quality Segment
Package Size
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing