
Valproic acid

Synonym(s):
2-Propylpentanoic acid, Valproic acid
Linear Formula:
(CH3CH2CH2)2CHCO2H
CAS Number:
Molecular Weight:
144.21
Products
Brand
Boiling Point (°C)
Formula Weight
Gene Alias
Manufacturer
Agency
Format
Quality Segment
Application
API Family
Reference Material Type
Special Grade
Technique