
Testosterone decanoate for system suitability

Synonym(s):
Testosterone decanoate, 17β-Hydroxy-4-androsten-3-one 17-decanoate, 4-Androsten-17β-ol-3-one 17-caprate
Empirical Formula (Hill Notation):
C29H46O3
CAS Number:
Molecular Weight:
442.67
Products
Formula Weight
Manufacturer
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing