
Steviol

Synonym(s):
(4α)-13-Hydroxykaur-16-en-18-oic acid, ent-13-Hydroxykauran-16-en-19-oic acid, NSC 226902
Empirical Formula (Hill Notation):
C20H30O3
CAS Number:
Molecular Weight:
318.45
Products
Formula Weight
Manufacturer
Physical Form
Product Line
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing