Skip to Content
Merck
Silica gel on TLC-PET foils with fluorescent indicator 254 nm, silica gel 60 matrix

Silica gel on TLC-PET foils

Silica gel on TLC-PET foils with fluorescent indicator 254 nm, silica gel 60 matrix
Synonym(s):
Poly TLC Plates: Silica w/o Fluorescence

Products

Product Category
Brand
Manufacturer
Length (cm)
Quality Segment
Fluorescence
Matrix Material
Support Material
Particle Size (µm)
Pore Size (Å)
Technique
Showing 1-1 of 1