
Quinoxyfen

Synonym(s):
(5,7-Dichloro-4-quinolyl) (4-fluorophenyl) ether
Empirical Formula (Hill Notation):
C15H8Cl2FNO
CAS Number:
Molecular Weight:
308.13
Products
Product Category
Brand
Formula Weight
Manufacturer
Product Line
Format
Quality Segment
Application
Reference Material Type
Special Grade
Technique
Showing 1-2 of 2