![Pivalic acid 99%](/deepweb/assets/sigmaaldrich/product/structures/126/852/e7d7ad66-bb5e-4838-8641-da17b00c4ff8/640/e7d7ad66-bb5e-4838-8641-da17b00c4ff8.png)
Pivalic acid
![Pivalic acid 99%](/deepweb/assets/sigmaaldrich/product/structures/126/852/e7d7ad66-bb5e-4838-8641-da17b00c4ff8/640/e7d7ad66-bb5e-4838-8641-da17b00c4ff8.png)
Synonym(s):
2,2-Dimethylpropionic acid, Trimethylacetic acid, Pivalic acid
Linear Formula:
(CH3)3CCOOH
CAS Number:
Molecular Weight:
102.13
Products
Product Category
Brand
Boiling Point (°C)
Formula Weight
Melting Point (°C)
Physical Form
Purity
Quality Segment
Functional Group
Markush Class
Markush Group
Reaction Type
Showing 1-2 of 2
Compare
Product Number
Description
Pricing