
Oxazepam for peak identification

Synonym(s):
Oxazepam, 7-Chloro-1,3-dihydro-3-hydroxy-5-phenyl-1,4(2H)-benzodiazepin-2-one
Empirical Formula (Hill Notation):
C15H11ClN2O2
CAS Number:
Molecular Weight:
286.71
Products
Formula Weight
Gene Alias
Manufacturer
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing