
N-Acetyltyrosine

Synonym(s):
N-Acetyl-L-tyrosine
Linear Formula:
4-(HO)C6H4CH2CH(NHCOCH3)CO2H
CAS Number:
Molecular Weight:
223.23
Products
Formula Weight
Manufacturer
Melting Point (°C)
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing