
N-Acetyl-L-tyrosine ethyl ester monohydrate

Synonym(s):
ATEE
Empirical Formula (Hill Notation):
C13H17NO4 · H2O
Products
Product Category
Brand
Color
Formula Weight
Physical Form
Optical Isomer
Amino Acid
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing