
Methyl (methylthio)acetate

Synonym(s):
Methyl (methylmercapto)acetate, Methyl S-methylthioglycolate
Linear Formula:
CH3SCH2COOCH3
Products
Product Category
Brand
Biological Source
Boiling Point (°C)
Formula Weight
Agency
Quality Segment
Application
Markush Class
Markush Group
Organoleptic Class
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing