
Methylene Blue

Synonym(s):
Methylthioninium chloride, Methylene blue
Empirical Formula (Hill Notation):
C16H18ClN3S
CAS Number:
Molecular Weight:
319.85
Products
Product Category
Brand
Color
Formula Weight
Physical Form
Research Area
Quality Segment
Showing 1-1 of 1
Compare
Product Number
Description
Pricing