
Lactobionic acid

Synonym(s):
4-O-β-D-Galactopyranosyl-D-gluconic acid
Empirical Formula (Hill Notation):
C12H22O12
CAS Number:
Molecular Weight:
358.30
Products
Product Category
Brand
Biological Source
Color
Formula Weight
Manufacturer
Melting Point (°C)
Physical Form
Purity
Format
Quality Segment
Application
Optical Isomer
API Family
Reference Material Type
Special Grade
Showing 1-3 of 3