
Hyoscine butylbromide

Synonym(s):
(−)-Scopolamine N-butyl bromide, (−)-N-Butylscopolamine bromide, Hyoscine N-butyl bromide
Empirical Formula (Hill Notation):
C21H30BrNO4
CAS Number:
Molecular Weight:
440.37
Products
Formula Weight
Manufacturer
Physical Form
Format
Application
API Family
Special Grade
Showing 1-2 of 2
Compare
Product Number
Description
Pricing