
Hydroxocobalamin acetate

Synonym(s):
Vitamin B12a acetate, Hydroxocobalamin acetate
Empirical Formula (Hill Notation):
C64H91CoN13O16P
CAS Number:
Molecular Weight:
1388.39
Products
Product Category
Brand
Biological Source
Color
Formula Weight
Manufacturer
Physical Form
Agency
Format
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade
Technique