![holo-Transferrin human ≥98%](/deepweb/assets/sigmaaldrich/product/images/299/235/8fe1255c-4632-4459-9a8b-bfd4f45b44be/640/8fe1255c-4632-4459-9a8b-bfd4f45b44be.jpg)
holo-Transferrin human
![holo-Transferrin human ≥98%](/deepweb/assets/sigmaaldrich/product/images/299/235/8fe1255c-4632-4459-9a8b-bfd4f45b44be/640/8fe1255c-4632-4459-9a8b-bfd4f45b44be.jpg)
Synonym(s):
Siderophilin, iron-saturated
CAS Number:
EC Number:
Products
Product Category
Brand
Biological Source
Gene Alias
Physical Form
Product Line
Purity
Tissue
Quality Segment
Technique
Showing 1-2 of 2
Compare
Product Number
Description
Pricing