Skip to Content
Merck
Fenthion oxon PESTANAL®, analytical standard

Fenthion oxon

Fenthion oxon PESTANAL®, analytical standard
Synonym(s):
O,O′-Dimethyl O′′-[3-methyl-4-(methylthio)phenyl] phosphate, Dimethyl 4-(methylthio)-m-tolyl phosphate, Fenoxon, Fenthoxon
Empirical Formula (Hill Notation):
C10H15O4PS
CAS Number:
Molecular Weight:
262.26

Products

Product Category
Brand
Formula Weight
Manufacturer
Product Line
Purity
Format
Quality Segment
Reference Material Type
Special Grade