
Dydrogesterone

Synonym(s):
9β,10α-Pregna-4,6-diene-3,20-dione
Empirical Formula (Hill Notation):
C21H28O2
CAS Number:
Molecular Weight:
312.45
Products
Formula Weight
Manufacturer
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing