
Dibenzo-30-crown-10

Synonym(s):
2,3,17,18-Dibenzo-1,4,7,10,13,16,19,22,25,28-decaoxacyclotriaconta-2,17-diene
Empirical Formula (Hill Notation):
C28H40O10
CAS Number:
Molecular Weight:
536.61
Products
Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Quality Segment
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing