
Coenzyme A sodium salt hydrate

Synonym(s):
CoA Na2
Empirical Formula (Hill Notation):
C21H36N7O16P3S · xNa+ · yH2O
Products
Product Category
Brand
Biological Source
Formula Weight
Physical Form
Product Line
Purity
Quality Segment
Technique
Showing 1-2 of 2
Compare
Product Number
Description
Pricing